4-(Aminomethyl)tetrahydropyran-4-ol ≥95%

Supplier: Matrix Scientific

074521-250MG 074521-5G
102536-854EA 640 USD
102536-854 102536-858
4-(Aminomethyl)tetrahydropyran-4-ol ≥95%
4-(Aminomethyl)tetrahydropyran-4-ol

Hazard: Irritant

Formula: C₆H₁₃NO₂
MDL Number: MFCD01628294
CAS Number: 783303-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR