2-Amino-5-bromo-4-chloro-3-nitropyridine ≥95%

Supplier: Matrix Scientific

074513-250MG 074513-5G
102536-808EA 1204.62 USD
102536-808 102536-812
2-Amino-5-bromo-4-chloro-3-nitropyridine ≥95%
2-Amino-5-bromo-4-chloro-3-nitropyridine

Hazard: Irritant

Formula: C₅H₃BrClN₃O₂
MDL Number: MFCD11110693
CAS Number: 942947-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR