4-Chloro-2-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

074510-250MG 074510-5G
102536-790EA 302.08 USD
102536-790 102536-794
4-Chloro-2-methoxypyrimidine ≥95%
4-Chloro-2-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₅ClN₂O
MW: 144.5590 g/mol
MDL Number: MFCD09025734
CAS Number: 51421-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR