2-Bromo-3,5-dichloroaniline ≥95%

Supplier: Matrix Scientific

074499-250MG 074499-5G
102536-732EA 640 USD
102536-732 102536-736
2-Bromo-3,5-dichloroaniline ≥95%
2-Bromo-3,5-dichloroaniline

Hazard: Irritant

Formula: C₆H₄BrCl₂N
MW: 240.9100 g/mol
MDL Number: MFCD09999825
CAS Number: 1211214-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR