(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulfonate ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-(+)-3-Aminotetrahydrofuran p-toluenesulphonate salt, (R)-(-)-3-aminotetrahydrofurantoluene-4-sulphonate, (R)-(+)-3-Aminotetrahydrofuran p-toluenesulphonate, (R)-Tetrahydrofuran-3-amine 4-methylbenzenesulphonate, (R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate
074492-1G
074492-10G
102536-694EA
253.85
USD
102536-694
102536-698
(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulfonate ≥95%
(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulfonate
Hazard: Irritant
Formula:
C₁₁H₁₇NO₄S MW: 259.32 g/mol |
MDL Number:
MFCD00040622 CAS Number: 111769-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...