Benzyl-3-hydroxypyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Benzyl 3-hydroxypyrrolidine-1-carboxylate

074481-1G 074481-10G
102536-650EA 284.62 USD
102536-650 102536-654
Benzyl-3-hydroxypyrrolidine-1-carboxylate ≥95%
Benzyl-3-hydroxypyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MW: 221.2500 g/mol
MDL Number: MFCD08061949
CAS Number: 95656-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR