4-Bromo-2-fluoro-5-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

074471-1G 074471-10G
102536-600EA 355.38 USD
102536-600 102536-604
4-Bromo-2-fluoro-5-methylbenzonitrile ≥95%
4-Bromo-2-fluoro-5-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₅BrFN
MW: 214.0300 g/mol
MDL Number: MFCD07782073
CAS Number: 916792-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR