3-Amino-5-nitrobenzylalkohol ≥95%
Supplier: Matrix Scientific
Synonyms:
(3-Amino-5-nitrophenyl)methanol
074465-250MG
074465-5G
102536-570EA
400
USD
102536-570
102536-574
3-Amino-5-nitrobenzylalkohol ≥95%
3-Amino-5-nitrobenzylalkohol
Hazard: Irritant
Formula:
C₇H₈N₂O₃ MW: 168.1500 g/mol |
MDL Number:
MFCD08236778 CAS Number: 90390-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...