6-Chloro-o-toluidine ≥95%

Supplier: Matrix Scientific

074448-100G 074448-25G
102536-490EA 250.77 USD
102536-490 102664-008
6-Chloro-o-toluidine ≥95%
6-Chloro-o-toluidine

Hazard: Toxic

Formula: C₇H₈ClN
Boiling Pt: 215 °C
Melting Pt: 2 °C
Density: 1.174
Flash Pt: 98 °C
MDL Number: MFCD00007679
CAS Number: 87-63-8
UN: 3429
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR