(S)-(-)-Tetrahydro-2-furoic acid ≥95%

Supplier: Matrix Scientific

074444-100G 074444-25G
102536-468EA 423.08 USD
102536-468 102664-006
(S)-(-)-Tetrahydro-2-furoic acid ≥95%
(S)-(-)-Tetrahydro-2-furoic acid

Hazard: Corrosive

Formula: C₅H₈O₃
Boiling Pt: 244 °C
Density: 1.213
MDL Number: MFCD00272639
CAS Number: 87392-07-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR