1-Boc-2-phenyl-4-piperidinone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-2-Phenyl-4-piperidinone
074429-250MG
074429-5G
102536-382EA
355.38
USD
102536-382
102536-386
1-Boc-2-phenyl-4-piperidinone ≥95%
1-Boc-2-phenyl-4-piperidinone
Hazard: Irritant
Formula:
C₁₆H₂₁NO₃ MW: 275.3400 g/mol |
MDL Number:
MFCD04035609 CAS Number: 849928-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...