5,6-Difluoroindoline-2,3-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6-Difluoroisatin

074422-1G 074422-10G
102536-342EA 533.85 USD
102536-342 102536-346
5,6-Difluoroindoline-2,3-dione ≥95%
5,6-Difluoroindoline-2,3-dione

Hazard: Irritant

Formula: C₈H₃F₂NO₂
MW: 183.110 g/mol
MDL Number: MFCD02998377
CAS Number: 774-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR