2-Phenylisobutyric acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-2-phenylpropanoic acid, α,α-Dimethylphenylacetic acid

074418-25G 074418-100G
102833-692EA 69.23 USD
102833-692 102833-694
2-Phenylisobutyric acid ≥95%
2-Phenylisobutyric acid

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
MDL Number: MFCD00014332
CAS Number: 826-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR