3-Fluoro-4-iodobenzoic acid ≥95%
Supplier: Matrix Scientific
074417-1G
074417-25G
102536-318EA
126.15
USD
102536-318
102664-002
3-Fluoro-4-iodobenzoic acid ≥95%
3-Fluoro-4-iodobenzoic acid
Hazard: Irritant
Formula:
C₇H₄FIO₂ MW: 266.0100 g/mol |
MDL Number:
MFCD07774250 CAS Number: 825-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...