4-Amino-3,6-dichloropyridazine ≥95%

Supplier: Matrix Scientific

074414-1G 074414-10G
102536-300EA 20 USD
102536-300 102536-304
4-Amino-3,6-dichloropyridazine ≥95%
4-Amino-3,6-dichloropyridazine

Hazard: Irritant

Formula: C₄H₃Cl₂N₃
MW: 163.9900 g/mol
MDL Number: MFCD01647253
CAS Number: 823-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR