2-Bromo-4-(trifluoromethyl)phenol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4-hydroxybenzotrifluoride

074412-5G 074412-25G
102536-290EA 113.85 USD
102536-290 102664-000
2-Bromo-4-(trifluoromethyl)phenol ≥95%
2-Bromo-4-(trifluoromethyl)phenol

Hazard: Irritant

Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
MDL Number: MFCD06657954
CAS Number: 81107-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR