1-Bromo-4-(trans-4-pentylcyclohexyl)benzene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-4-(trans-4-N-pentylcyclohexyl)benzene

074405-1G 074405-10G
102536-254EA 49.23 USD
102536-254 102536-258
1-Bromo-4-(trans-4-pentylcyclohexyl)benzene ≥95%
1-Bromo-4-(trans-4-pentylcyclohexyl)benzene

Hazard: Irritant

Formula: C₁₇H₂₅Br
MW: 309.2800 g/mol
MDL Number: MFCD00577359
CAS Number: 79832-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR