endo-3-Amine-9-methyl-9-azabicyclo[3,3,1]nonane ≥95%

Supplier: Matrix Scientific

Synonyms: Granisetron impurity E

074391-5G 074391-25G
102536-186EA 118.46 USD
102536-186 102833-690
endo-3-Amine-9-methyl-9-azabicyclo[3,3,1]nonane ≥95%
endo-3-Amine-9-methyl-9-azabicyclo[3,3,1]nonane

Hazard: Irritant

Formula: C₉H₁₈N₂
MW: 154.25 g/mol
MDL Number: MFCD07374273
CAS Number: 76272-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR