(1-Methylpiperidin-4-yl)methanamine ≥95%

Supplier: Matrix Scientific

074379-1G 074379-10G
102536-120EA 109.23 USD
102536-120 102536-124
(1-Methylpiperidin-4-yl)methanamine ≥95%
(1-Methylpiperidin-4-yl)methanamine

Hazard: Corrosive

Formula: C₇H₁₆N₂
MDL Number: MFCD05022430
CAS Number: 7149-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR