3-Bromo-5-fluoroaniline ≥95%

Supplier: Matrix Scientific

074376-5G 074376-25G
102536-106EA 369.23 USD
102536-106 102833-688
3-Bromo-5-fluoroaniline ≥95%
3-Bromo-5-fluoroaniline

Hazard: Toxic

Formula: C₆H₅BrFN
MDL Number: MFCD07779529
CAS Number: 134168-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR