2,6-Dibromo-p-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3,5-dibromotoluene, 2,6-Dibromo-4-methylaniline

074370-5G 074370-100G
102536-072EA 36.92 USD
102536-072 102536-076
2,6-Dibromo-p-toluidine ≥95%
2,6-Dibromo-p-toluidine

Hazard: Irritant

Formula: C₇H₇Br₂N
MW: 264.95 g/mol
MDL Number: MFCD00007641
CAS Number: 6968-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR