3-Aminocyclohexanol ≥95%
Supplier: Matrix Scientific
074367-1G
074367-25G
102536-056EA
107.69
USD
102536-056
102551-890
3-Aminocyclohexanol ≥95%
3-Aminocyclohexanol
Hazard: Irritant-Harmful/Refrigerate/Stored under Argon
Formula:
C₆H₁₃NO MW: 115.1700 g/mol Melting Pt: 64…69 °C |
MDL Number:
MFCD03790923 CAS Number: 6850-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...