(R)-(+)-3-Amino-1,2-propanediol ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-3-Aminopropane-1,2-diol, (R)-3-Amino-1,2-propanediol

074361-5G 074361-100G
102536-022EA 198.46 USD
102536-022 102536-026
(R)-(+)-3-Amino-1,2-propanediol ≥95%
(R)-(+)-3-Amino-1,2-propanediol

Hazard: Irritant

Formula: C₃H₉NO₂
MW: 91.11 g/mol
MDL Number: MFCD00798261
CAS Number: 66211-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR