4-Methyl-3-nitrobenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-2-nitro-4-(trifluoromethyl)benzene

074359-1G 074359-10G
102536-014EA 233.85 USD
102536-014 102536-018
4-Methyl-3-nitrobenzotrifluoride ≥95%
4-Methyl-3-nitrobenzotrifluoride

Hazard: Irritant

Formula: C₈H₆F₃NO₂
MW: 205.1300 g/mol
MDL Number: MFCD00272402
CAS Number: 65754-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR