(2S)-2-Amino-2-phenylethanamide ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-Amino-2-phenylacetamide, (S)-(+)-2-Phenylglycine Amide

074356-5G 074356-25G
102535-998EA 21.54 USD
102535-998 102833-686
(2S)-2-Amino-2-phenylethanamide ≥95%
(2S)-2-Amino-2-phenylethanamide

Hazard: Irritant

Formula: C₈H₁₀N₂O
MW: 150.1800 g/mol
MDL Number: MFCD03791027
CAS Number: 6485-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR