4-((Dimethylamino)methyl)aniline ≥95%

Supplier: Matrix Scientific

074353-1G 074353-10G
102535-978EA 109.23 USD
102535-978 102535-982
4-((Dimethylamino)methyl)aniline ≥95%
4-((Dimethylamino)methyl)aniline

Hazard: Irritant

Formula: C₉H₁₄N₂
MDL Number: MFCD00035953
CAS Number: 6406-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR