2-Iodo-4-nitroaniline ≥95%

Supplier: Matrix Scientific

074349-1G 074349-10G
102535-960EA 178.46 USD
102535-960 102535-964
2-Iodo-4-nitroaniline ≥95%
2-Iodo-4-nitroaniline

Hazard: Irritant

Formula: C₆H₅IN₂O₂
MW: 264.02 g/mol
MDL Number: MFCD00955758
CAS Number: 6293-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR