1-Benzylpiperidine-4-carbonitrile ≥95%

Supplier: Matrix Scientific

074344-1G 074344-10G
102535-932EA 533.85 USD
102535-932 102535-936
1-Benzylpiperidine-4-carbonitrile ≥95%
1-Benzylpiperidine-4-carbonitrile

Hazard: Irritant

Formula: C₁₃H₁₆N₂
MW: 200.2795 g/mol
MDL Number: MFCD01540574
CAS Number: 62718-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR