2-Ethoxy-5-pyridineboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Ethoxy-5-pyridylboronic acid, 6-Ethoxy-3-pyridylboronic acid, 2-Ethoxypyridine-5-boronic acid, (6-Ethoxypyridin-3-yl)boronic acid, 6-Ethoxypyridine-3-boronic acid

074334-1G 074334-10G
102535-880EA 23.08 USD
102535-880 102535-884
2-Ethoxy-5-pyridineboronic acid ≥95%
2-Ethoxy-5-pyridineboronic acid

Hazard: Irritant

Formula: C₇H₁₀BNO₃
MW: 166.97 g/mol
MDL Number: MFCD06201032
CAS Number: 612845-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR