2-Hydroxycyclohexanecarboxylic acid ≥95%
Supplier: Matrix Scientific
074332-250MG
074332-5G
102535-868EA
412.31
USD
102535-868
102535-872
2-Hydroxycyclohexanecarboxylic acid ≥95%
2-Hydroxycyclohexanecarboxylic acid
Hazard: Irritant
Formula:
C₇H₁₂O₃ |
MDL Number:
MFCD02684437 CAS Number: 609-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...