2-Nitro-m-toluidine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methyl-2-nitroaniline
074328-5G
074328-25G
102535-852EA
90.77
USD
102535-852
102833-684
2-Nitro-m-toluidine ≥95%
2-Nitro-m-toluidine
Hazard: Toxic/Refrigerate
Formula:
C₇H₈N₂O₂ MW: 152.1500 g/mol Melting Pt: 105°C |
MDL Number:
MFCD00074851 CAS Number: 601-87-6 UN: 2660 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...