2-Bromo-5-methoxyaniline ≥95%
Supplier: Matrix Scientific
074326-5G
074326-25G
102535-842EA
26.15
USD
102535-842
102551-888
2-Bromo-5-methoxyaniline ≥95%
2-Bromo-5-methoxyaniline
Hazard: Irritant-Harmful/Refrigerate
Formula:
C₇H₈BrNO |
MDL Number:
MFCD00070741 CAS Number: 59557-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...