4-(Trifluoromethyl)quinoline-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

074322-250MG 074322-5G
102535-820EA 400 USD
102535-820 102535-824
4-(Trifluoromethyl)quinoline-2-carboxylic acid ≥95%
4-(Trifluoromethyl)quinoline-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₆F₃NO₂
MW: 241.17
MDL Number: MFCD06796588
CAS Number: 588702-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR