2,6-Dimethyl-4-hydroxybenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-2,6-dimethylbenzonitrile

074320-1G 074320-10G
102535-808EA 107.69 USD
102535-808 102535-812
2,6-Dimethyl-4-hydroxybenzonitrile ≥95%
2,6-Dimethyl-4-hydroxybenzonitrile

Hazard: Irritant

Formula: C₉H₉NO
MDL Number: MFCD00238591
CAS Number: 58537-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR