5-Bromo-2-chloro-4-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

074310-1G 074310-10G
102535-756EA 15.38 USD
102535-756 102535-760
5-Bromo-2-chloro-4-methoxypyrimidine ≥95%
5-Bromo-2-chloro-4-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₄BrClN₂O
MW: 223.4600 g/mol
MDL Number: MFCD03001373
CAS Number: 57054-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR