2-Chloro-4-(trifluoromethyl)benzyl alcohol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(trifluoromethyl)benzylalcohol

074302-1G 074302-10G
102535-714EA 43.08 USD
102535-714 102535-718
2-Chloro-4-(trifluoromethyl)benzyl alcohol ≥95%
2-Chloro-4-(trifluoromethyl)benzyl alcohol

Hazard: Irritant

Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
MDL Number: MFCD04972757
CAS Number: 56456-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR