2-Amino-3-chlorobenzonitrile ≥95%
Supplier: Matrix Scientific
074291-1G
074291-10G
102535-658EA
178.46
USD
102535-658
102535-662
2-Amino-3-chlorobenzonitrile ≥95%
2-Amino-3-chlorobenzonitrile
Hazard: Irritant
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol |
MDL Number:
MFCD03425881 CAS Number: 53312-77-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...