3-Methoxy-2-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

074288-1G 074288-10G
102535-642EA 178.46 USD
102535-642 102535-646
3-Methoxy-2-nitrobenzaldehyde ≥95%
3-Methoxy-2-nitrobenzaldehyde

Hazard: Irritant

Formula: C₈H₇NO₄
MDL Number: MFCD00007135
CAS Number: 53055-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR