2-(3,5-Dichlorophenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

074284-5G 074284-25G
102535-624EA 67.69 USD
102535-624 102663-994
2-(3,5-Dichlorophenyl)acetonitrile ≥95%
2-(3,5-Dichlorophenyl)acetonitrile

Hazard: Irritant

Formula: C₈H₅Cl₂N
MW: 186.0380 g/mol
MDL Number: MFCD09923674
CAS Number: 52516-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR