5-Fluoro-m-toluidine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-5-methylaniline
074281-1G
074281-10G
102535-604EA
161.54
USD
102535-604
102535-608
5-Fluoro-m-toluidine ≥95%
5-Fluoro-m-toluidine
Hazard: Irritant
Formula:
C₇H₈FN MW: 125.14 g/mol |
MDL Number:
MFCD00077538 CAS Number: 52215-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...