Methyl-5-(tert-butyl)isoxazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-(tert-butyl)isoxazole-3-carboxylate

074279-250MG 074279-5G
102535-592EA 427.69 USD
102535-592 102535-596
Methyl-5-(tert-butyl)isoxazole-3-carboxylate ≥95%
Methyl-5-(tert-butyl)isoxazole-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₃NO₃
MW: 183.2000 g/mol
MDL Number: MFCD03778975
CAS Number: 517870-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR