3-Chloro-2-iodotoluene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-iodo-3-methylbenzene

074275-5G 074275-10G
102535-574EA 307.69 USD
102535-574 102535-576
3-Chloro-2-iodotoluene ≥95%
3-Chloro-2-iodotoluene

Hazard: Irritant

Formula: C₇H₆ClI
MW: 252.48 g/mol
MDL Number: MFCD00051756
CAS Number: 5100-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR