4-Bromo-2,6-dimethylpyridine ≥95%

Supplier: Matrix Scientific

074274-5G 074274-25G
102535-568EA 96.92 USD
102535-568 102663-992
4-Bromo-2,6-dimethylpyridine ≥95%
4-Bromo-2,6-dimethylpyridine

Hazard: Irritant

Formula: C₇H₈BrN
Melting Pt: 32…34 °C
MDL Number: MFCD00671475
CAS Number: 5093-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR