5-Methoxy-o-toluidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-methoxytoluene, 3-Amino-4-methylanisole, 1-Amino-3-methoxy-6-methylbenzene, 6-Methyl-m-anisidine, 5-Methoxy-2-methylaniline

074273-25G 074273-100G
102663-988EA 558.46 USD
102663-988 102663-990
5-Methoxy-o-toluidine ≥95%
5-Methoxy-o-toluidine

Hazard: Irritant

Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00075057
CAS Number: 50868-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR