2,6-Dichlorophenylhydrazinium chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dichlorophenylhydrazine hydrochloride, 1-(2,6-Dichlorophenyl)hydrazine hydrochloride

074272-5G 074272-100G
102535-554EA 143.08 USD
102535-554 102535-558
2,6-Dichlorophenylhydrazinium chloride ≥95%
2,6-Dichlorophenylhydrazinium chloride

Hazard: Irritant

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00012930
CAS Number: 50709-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR