(2-Chloro-5-fluorophenyl)hydrazine hydrochloride ≥95%

Supplier: Matrix Scientific

074265-1G 074265-10G
102535-518EA 233.85 USD
102535-518 102535-522
(2-Chloro-5-fluorophenyl)hydrazine hydrochloride ≥95%
(2-Chloro-5-fluorophenyl)hydrazine hydrochloride

Hazard: Irritant

Formula: C₆H₇Cl₂FN₂
MDL Number: MFCD03094167
CAS Number: 502496-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR