7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-chromen-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Methoxy-8-(3-methyl-2-butenyl)coumarin, 7-Methoxy-8-(3-methylbut-2-en-1-yl)-2H-chromen-2-one, Osthole

074257-1G 074257-10G
102535-478EA 355.38 USD
102535-478 102535-482
7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-chromen-2-one ≥95%
7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-chromen-2-one

Hazard: Irritant

Formula: C₁₅H₁₆O₃
MW: 244.2857 g/mol
MDL Number: MFCD00076049
CAS Number: 484-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR