2-Fluoro-p-anisidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-anisidine, 2-Fluoro-4-methoxyaniline, 4-Amino-3-fluoroanisole

074248-5G 074248-25G
102535-432EA 304.62 USD
102535-432 102663-986
2-Fluoro-p-anisidine ≥95%
2-Fluoro-p-anisidine

Hazard: Irritant

Formula: C₇H₈FNO
Melting Pt: 50°C
MDL Number: MFCD00047827
CAS Number: 458-52-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR