6-Methylanthranil acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-methylbenzoic acid

074242-5G 074242-25G
102535-398EA 26.15 USD
102535-398 102551-882
6-Methylanthranil acid ≥95%
6-Methylanthranil acid

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00007809
CAS Number: 4389-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR