2-Bromo-p-anisaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-methoxybenzaldehyde

074240-1G 074240-25G
102535-388EA 38.46 USD
102535-388 102663-984
2-Bromo-p-anisaldehyde ≥95%
2-Bromo-p-anisaldehyde

Hazard: Irritant

Formula: C₈H₇BrO₂
MW: 215.04 g/mol
MDL Number: MFCD09788636
CAS Number: 43192-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR